(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118219-1G 118219-500MG 118219-5G 118219-25G
77464-148EA 415.09 USD
77464-148 77476-940 77487-020 77497-868
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(phenyl)methanol ≥95%
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₉H₁₄BrFO
MW: 357.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783971
CAS Number: 2918843-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR