(2-Fluoro-5-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118205-1G 118205-500MG 118205-5G 118205-25G
77464-144EA 415.09 USD
77464-144 77476-936 77487-016 77497-866
(2-Fluoro-5-methylphenyl)(phenyl)methanol ≥95%
(2-Fluoro-5-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃FO
MW: 216.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16783385
CAS Number: 1250965-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR