(5-Chloro-2-fluoro-3-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118201-1G 118201-500MG 118201-5G 118201-25G
77464-140EA 415.09 USD
77464-140 77476-932 77487-012 77497-862
(5-Chloro-2-fluoro-3-methylphenyl)(phenyl)methanol ≥95%
(5-Chloro-2-fluoro-3-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂ClFO
MW: 250.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783956
CAS Number: 2918933-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR