(4-Bromo-3-chloro-2-fluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118190-1G 118190-500MG 118190-5G 118190-25G
77464-136EA 415.09 USD
77464-136 77476-928 77487-008 77497-858
(4-Bromo-3-chloro-2-fluorophenyl)(phenyl)methanol ≥95%
(4-Bromo-3-chloro-2-fluorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₉BrClFO
MW: 315.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783946
CAS Number: 1978667-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR