(2-Bromo-5-fluoropyridin-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118172-1G 118172-500MG 118172-5G 118172-25G
77464-132EA 415.09 USD
77464-132 77476-924 77487-004 77497-854
(2-Bromo-5-fluoropyridin-4-yl)(phenyl)methanol ≥95%
(2-Bromo-5-fluoropyridin-4-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₉BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34783928
CAS Number: 1114523-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR