(2-Bromo-6-fluoro-4-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118161-1G 118161-500MG 118161-5G 118161-25G
77464-128EA 415.09 USD
77464-128 77476-920 77487-000 77497-852
(2-Bromo-6-fluoro-4-methylphenyl)(phenyl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783917
CAS Number: 2918931-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR