2-Chloro-1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

117736-1G 117736-500MG 117736-5G
77464-124EA 696.28 USD
77464-124 77476-916 77486-996
2-Chloro-1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃Cl₂F₃O
MW: 243.0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823509
CAS Number: 1850602-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR