1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117469-1G 117469-500MG 117469-5G 117469-25G
77464-120EA 415.09 USD
77464-120 77476-912 77486-992 77497-842
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFOS
MW: 293.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783502
CAS Number: 2918892-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR