1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117457-1G 117457-500MG 117457-5G 117457-25G
77464-116EA 415.09 USD
77464-116 77476-908 77486-988 77497-840
1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₉H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34783491
CAS Number: 2918845-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR