1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117445-1G 117445-500MG 117445-5G 117445-25G
77464-112EA 415.09 USD
77464-112 77476-904 77486-984 77497-836
1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-6-fluoro-3-propoxyphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₁₈BrFO₂
MW: 305.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783481
CAS Number: 2918880-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR