Ethyl 2-(3-bromo-2-fluoro-5-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

117374-1G 117374-500MG 117374-5G
77464-104EA 646.84 USD
77464-104 77476-896 77486-976
Ethyl 2-(3-bromo-2-fluoro-5-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-2-fluoro-5-methylphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₁H₁₀BrFO₃
MW: 289.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823473
CAS Number: 1898050-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR