1-(3-Bromofuran-2-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

117290-1G 117290-500MG 117290-5G 117290-25G
77464-100EA 426.42 USD
77464-100 77476-892 77486-972 77497-828
1-(3-Bromofuran-2-yl)-2,2-difluoroethanone ≥97%
1-(3-Bromofuran-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₆H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26045321
CAS Number: 1566884-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR