1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

117015-1G 117015-500MG 117015-5G 117015-25G
77464-096EA 376.98 USD
77464-096 77476-888 77486-966 77497-822
1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanol ≥95%
1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanol

Precaución: For research use only

Formula: C₈H₅BrF₄O
MW: 273.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16067891
CAS Number: 1039827-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR