1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

117010-1G 117010-500MG 117010-5G
77464-092EA 696.28 USD
77464-092 77476-884 77486-962
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanol ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanol

Precaución: For research use only

Formula: C₈H₄BrClF₄O
MW: 307.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29913453
CAS Number: 943830-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR