4-Bromo-2-chloro-3-fluoroiodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-chloro-2-fluoro-4-iodobenzene

116927-1G 116927-500MG 116927-5G 116927-25G
77464-088EA 97.85 USD
77464-088 77476-880 77486-958 77497-816
4-Bromo-2-chloro-3-fluoroiodobenzene ≥95%
4-Bromo-2-chloro-3-fluoroiodobenzene

Precaución: For research use only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30183258
CAS Number: 1917307-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR