(3-Bromophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

116710-1G 116710-500MG 116710-5G 116710-25G
77464-084EA 203.94 USD
77464-084 77476-876 77486-954 77497-810
(3-Bromophenyl)(phenyl)methanol ≥95%
(3-Bromophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00449886
CAS Number: 63012-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR