(3-Bromo-2-fluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

116693-1G 116693-500MG 116693-5G
77464-080EA 505.73 USD
77464-080 77476-872 77486-950
(3-Bromo-2-fluorophenyl)(phenyl)methanol ≥95%
(3-Bromo-2-fluorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD25541941
CAS Number: 1428881-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR