(2-Ethoxy-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

116657-1G 116657-500MG 116657-5G 116657-25G
77464-076EA 415.09 USD
77464-076 77476-868 77486-946 77497-806
(2-Ethoxy-3-methylphenyl)(methyl)sulfane ≥95%
(2-Ethoxy-3-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783211
CAS Number: 2918855-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR