2-Ethoxy-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

116652-1G 116652-500MG 116652-5G 116652-25G
77464-072EA 173.04 USD
77464-072 77476-864 77486-942 77497-802
2-Ethoxy-3-methylbenzoic acid ≥95%
2-Ethoxy-3-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12804771
CAS Number: 103324-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR