1,3-Diethyl-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

116495-1G 116495-500MG 116495-5G 116495-25G
77464-068EA 100.94 USD
77464-068 77476-860 77486-938 77497-800
1,3-Diethyl-2-iodobenzene ≥95%
1,3-Diethyl-2-iodobenzene

Precaución: For research use only

Formula: C₁₀H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD09878114
CAS Number: 92015-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR