2,4-Dimethoxy-3-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

116401-1G 116401-500MG 116401-5G 116401-25G
77464-060EA 415.09 USD
77464-060 77476-852 77486-930 77497-792
2,4-Dimethoxy-3-(methoxymethoxy)benzaldehyde ≥97%
2,4-Dimethoxy-3-(methoxymethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₅
MW: 226.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783168
CAS Number: 2901110-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR