1-(2-Chloro-6-fluoro-3-methylphenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

116156-1G 116156-500MG 116156-5G 116156-25G
77464-056EA 415.09 USD
77464-056 77476-848 77486-926 77497-790
1-(2-Chloro-6-fluoro-3-methylphenyl)-2-fluoroethanone ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₉H₇ClF₂O
MW: 204.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32869201
CAS Number: 2918910-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR