3-Bromo-4-fluoro-5-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

115615-1G 115615-500MG 115615-5G 115615-25G
77464-052EA 368.74 USD
77464-052 77476-844 77486-922 77497-784
3-Bromo-4-fluoro-5-hydroxybenzoic acid ≥95%
3-Bromo-4-fluoro-5-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₄BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD25978048
CAS Number: 1628539-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR