1-(4-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115514-1G 115514-500MG 115514-5G 115514-25G
77464-048EA 226.6 USD
77464-048 77476-840 77486-918 77497-778
1-(4-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol ≥97%
1-(4-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD26686464
CAS Number: 1537970-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR