1-(3-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol ≥96%

Supplier: AOBChem USA

Warning

115502-1G 115502-500MG 115502-5G
77464-044EA 447.02 USD
77464-044 77476-836 77486-914
1-(3-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol ≥96%
1-(3-Bromo-2,6-difluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34823103
CAS Number: 1542333-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR