1-(2-Bromo-6-fluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115497-1G 115497-500MG 115497-5G 115497-25G
77464-040EA 427.45 USD
77464-040 77476-832 77486-910 77497-774
1-(2-Bromo-6-fluorophenyl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-6-fluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD26138240
CAS Number: 1520149-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR