1-(3-Methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115468-1G 115468-500MG 115468-5G 115468-25G
77464-036EA 365.65 USD
77464-036 77476-828 77486-906 77497-770
1-(3-Methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD12153725
CAS Number: 32578-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR