1-(3-Methoxyphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115466-1G 115466-500MG 115466-5G 115466-25G
77464-032EA 305.91 USD
77464-032 77476-824 77486-902 77497-766
1-(3-Methoxyphenyl)-2-methylpropan-1-ol ≥97%
1-(3-Methoxyphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153457
CAS Number: 61751-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR