1-(3-Bromophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115458-1G 115458-500MG 115458-5G
77464-028EA 471.74 USD
77464-028 77476-820 77486-898
1-(3-Bromophenyl)-2-methylpropan-1-ol ≥97%
1-(3-Bromophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153602
CAS Number: 186000-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR