1-(4-Methoxyphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

115451-1G 115451-500MG 115451-5G 115451-25G
77464-024EA 207.03 USD
77464-024 77476-816 77486-894 77497-760
1-(4-Methoxyphenyl)-2-methylpropan-1-one ≥95%
1-(4-Methoxyphenyl)-2-methylpropan-1-one

Precaución: For research use only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00095003
CAS Number: 2040-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR