2,3,4-Trifluoro-5-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

114618-1G 114618-5G 114618-25G
77464-016EA 71.07 USD
77464-016 77486-882 77497-742
2,3,4-Trifluoro-5-methoxybenzoic acid ≥95%
2,3,4-Trifluoro-5-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD11973916
CAS Number: 38233-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR