2-(2-(Benzyloxy)-5-bromophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

114014-1G 114014-500MG 114014-5G 114014-25G
77464-012EA 415.09 USD
77464-012 77476-808 77486-878 77497-734
2-(2-(Benzyloxy)-5-bromophenyl)-1,3-dioxolane ≥97%
2-(2-(Benzyloxy)-5-bromophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₆H₁₅BrO₃
MW: 335.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34782070
CAS Number: 177759-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR