5-acetyl-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

113986-1G 113986-500MG 113986-5G
77464-008EA 437.75 USD
77464-008 77476-804 77486-874
5-acetyl-2-methylbenzoic acid ≥97%
5-acetyl-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14536116
CAS Number: 1216048-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR