4-Bromo-6-chloro-2-fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

113728-1G 113728-500MG 113728-5G 113728-25G
77464-004EA 415.09 USD
77464-004 77476-800 77486-870 77497-728
4-Bromo-6-chloro-2-fluoro-3-hydroxybenzaldehyde ≥97%
4-Bromo-6-chloro-2-fluoro-3-hydroxybenzaldehyde

Precaución: For research use only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34822794
CAS Number: 2969453-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR