5-(4-Bromophenyl)pyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

113442-1G 113442-500MG 113442-5G 113442-25G
77464-000EA 346.08 USD
77464-000 77476-796 77486-866 77497-724
5-(4-Bromophenyl)pyrimidin-2-amine ≥97%
5-(4-Bromophenyl)pyrimidin-2-amine

Precaución: For research use only

Formula: C₁₀H₈BrN₃
MW: 250.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072508
CAS Number: 31483-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR