(4-(tert-Butyl)-2-isopropoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

112844-1G 112844-500MG 112844-5G 112844-25G
77463-996EA 415.09 USD
77463-996 77476-792 77486-858 77497-716
(4-(tert-Butyl)-2-isopropoxyphenyl)(methyl)sulfane ≥97%
(4-(tert-Butyl)-2-isopropoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₄H₂₂OS
Storage Temperature: Ambient
MDL Number: MFCD34822438
CAS Number: 2969341-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR