(4-(tert-Butyl)-2-ethoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

112833-1G 112833-500MG 112833-5G 112833-25G
77463-992EA 415.09 USD
77463-992 77476-788 77486-854 77497-712
(4-(tert-Butyl)-2-ethoxyphenyl)(methyl)sulfane ≥95%
(4-(tert-Butyl)-2-ethoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34781687
CAS Number: 2918912-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR