4-(tert-Butyl)-2-ethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

112830-1G 112830-500MG 112830-5G 112830-25G
77463-988EA 340.93 USD
77463-988 77476-786 77486-850 77497-708
4-(tert-Butyl)-2-ethoxybenzaldehyde ≥95%
4-(tert-Butyl)-2-ethoxybenzaldehyde

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23160063
CAS Number: 1073145-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR