(4-(Ethylthio)-2,3-difluorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

112440-1G 112440-500MG 112440-5G 112440-25G
77463-984EA 415.09 USD
77463-984 77476-782 77486-846 77497-702
(4-(Ethylthio)-2,3-difluorophenyl)boronic acid ≥95%
(4-(Ethylthio)-2,3-difluorophenyl)boronic acid

Precaución: For research use only

Formula: C₈H₉BF₂O₂S
MW: 218.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34781545
CAS Number: 2340233-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR