1-Bromo-4-chloro-2-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

111400-1G 111400-5G 111400-25G
77463-980EA 60.77 USD
77463-980 77486-842 77497-698
1-Bromo-4-chloro-2-propoxybenzene ≥95%
1-Bromo-4-chloro-2-propoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043813
CAS Number: 1881331-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR