4-Bromo-2,3-dimethylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

111236-1G 111236-500MG 111236-5G
77463-976EA 524.27 USD
77463-976 77476-776 77486-838
4-Bromo-2,3-dimethylbenzonitrile ≥95%
4-Bromo-2,3-dimethylbenzonitrile

Precaución: For research use only

Formula: C₉H₈BrN
MW: 210.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917143
CAS Number: 1349716-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR