5-(furan-2-yl)pyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

111202-1G 111202-500MG 111202-5G 111202-25G
77463-972EA 423.33 USD
77463-972 77476-772 77486-834 77497-688
5-(furan-2-yl)pyrimidin-2-amine ≥97%
5-(furan-2-yl)pyrimidin-2-amine

Precaución: For research use only

Formula: C₈H₇N₃O
MW: 161.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877092
CAS Number: 1111104-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR