5-Hydroxy-2-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

11098-1G 11098-500MG 11098-5G 11098-25G
77463-968EA 144.2 USD
77463-968 77476-768 77486-830 77497-684
5-Hydroxy-2-(trifluoromethoxy)benzoic acid ≥95%
5-Hydroxy-2-(trifluoromethoxy)benzoic acid

Precaución: For research use only

Formula: C₈H₅F₃O₄
MW: 222.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397465
CAS Number: 851341-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR