2,4-Dimethoxy-3-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

110767-1G 110767-500MG 110767-5G 110767-25G
77463-960EA 415.09 USD
77463-960 77476-762 77486-820 77497-674
2,4-Dimethoxy-3-(methylthio)pyridine ≥95%
2,4-Dimethoxy-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780795
CAS Number: 2918904-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR