(4-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥95%

Supplier: AOBChem USA

Warning

110663-1G 110663-500MG 110663-5G 110663-25G
77463-956EA 415.09 USD
77463-956 77476-758 77486-816 77497-670
(4-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥95%
(4-Chloro-2-fluoro-1,3-phenylene)bis(methylsulfane)

Precaución: For research use only

Formula: C₈H₈ClFS₂
MW: 222.731 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780744
CAS Number: 2918843-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR