2-Ethoxy-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

109751-1G 109751-500MG 109751-5G 109751-25G
77463-948EA 415.09 USD
77463-948 77476-750 77486-808 77497-656
2-Ethoxy-6-(methylthio)phenol ≥97%
2-Ethoxy-6-(methylthio)phenol

Precaución: For research use only

Formula: C₉H₁₂O₂S
MW: 184.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34821781
CAS Number: 2918958-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR