2-(3-Methyl-5-(methylthio)phenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

109560-1G 109560-500MG 109560-5G 109560-25G
77463-944EA 415.09 USD
77463-944 77476-746 77486-804 77497-652
2-(3-Methyl-5-(methylthio)phenyl)propan-2-ol ≥97%
2-(3-Methyl-5-(methylthio)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34821768
CAS Number: 2918963-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR