2-Chloro-4-methoxy-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

109357-1G 109357-500MG 109357-5G 109357-25G
77463-940EA 415.09 USD
77463-940 77476-742 77486-800 77497-644
2-Chloro-4-methoxy-3-(methylthio)pyridine ≥97%
2-Chloro-4-methoxy-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₇H₈ClNOS
Storage Temperature: Ambient
MDL Number: MFCD34780130
CAS Number: 2901083-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR