4-Bromo-5-chloro-2-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

108894-1G 108894-500MG 108894-5G 108894-25G
77463-936EA 230.72 USD
77463-936 77476-738 77486-796 77497-638
4-Bromo-5-chloro-2-methoxypyridine ≥95%
4-Bromo-5-chloro-2-methoxypyridine

Precaución: For research use only

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609882
CAS Number: 1211534-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR