4-Bromo-3,5-difluoropyridine ≥97%

Supplier: AOBChem USA

Warning

108872-1G 108872-5G 108872-25G
77463-932EA 64.89 USD
77463-932 77486-792 77497-634
4-Bromo-3,5-difluoropyridine ≥97%
4-Bromo-3,5-difluoropyridine

Precaución: For research use only

Formula: C₅H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD11505879
CAS Number: 1092352-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR